Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM102 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570151
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570151 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570151 Supplier Invitrogen™ Supplier No. PA570151
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 93%; Dog: 100%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 93%.

Selectively involved in CSF2 deprivation-induced apoptosis via a mitochondria-dependent pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM102
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TMEM102
Gene Accession No. Q3UPR7
Gene Alias 4632403M07Rik; Cbap; common beta-chain associated protein; common beta-chain-associated protein; LOW QUALITY PROTEIN: transmembrane protein 102; RGD1559450; TMEM102; Tmem102-ps; transmembrane protein 102; transmembrane protein 102, pseudogene
Gene Symbols TMEM102
Host Species Rabbit
Immunogen synthetic peptide corresponding to a region of mouse TMEM102 (aa 64-113).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 380705
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.