Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TMEM106B Mouse anti-Human, Clone: 5D1F8, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
| Antigen | TMEM106B |
|---|---|
| Clone | 5D1F8 |
| Concentration | 1.48 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6420
![]()
|
Proteintech Group Inc
603331IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Transmembrane protein 106B (TMEM106B) is a single-pass transmembrane protein that is thought to be a novel risk factor for frontotemporal lobar degeneration (FTLD), a group of clinically, pathologically and genetically heterogeneous disorders associated with atrophy in the frontal lobe and temporal lobe of the brain. The actual role of TMEM106B, and that of the closely related protein TMEM106A are still undetermined.Specifications
| TMEM106B | |
| 1.48 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| TMEM106B, transmembrane protein 106B | |
| TMEM106B | |
| IgG1 | |
| Protein G | |
| Antibody |
| 5D1F8 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q9NUM4 | |
| 54664 | |
| TMEM106B Fusion Protein Ag21448 | |
| Primary | |
| -20°C | |
| TMEM106B |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title