Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TMEM106B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5100353
Description
Antibody detects endogenous levels of total TMEM106B.
Transmembrane protein 106B (TMEM106B) is a single-pass transmembrane protein that is thought to be a novel risk factor for frontotemporal lobar degeneration (FTLD), a group of clinically, pathologically and genetically heterogeneous disorders associated with atrophy in the frontal lobe and temporal lobe of the brain. The actual role of TMEM106B, and that of the closely related protein TMEM106A are still undetermined.
Specifications
TMEM106B | |
Polyclonal | |
Unconjugated | |
TMEM106B | |
2310036D22Rik; 5830455K21Rik; 6430519M21Rik; AI428776; AI661344; hypothetical protein LOC508903; LRRGT00101; tmem106b; tmem106bb; Transmembrane protein 106B; transmembrane protein 106Bb; zgc:114147 | |
Rabbit | |
Affinity chromatography | |
RUO | |
54664, 71900 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q80X71, Q9NUM4 | |
TMEM106B | |
A synthesized peptide derived from human TMEM106B(Accession Q9NUM4), corresponding to amino acid residues V45-K95. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction