Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM106B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534353
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534353 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534353 Supplier Invitrogen™ Supplier No. PA534353
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is human liver tissue lysate. PA5-34353 can be used with blocking peptide PEP-1396.

Transmembrane protein 106B (TMEM106B) is a single-pass transmembrane protein that is thought to be a novel risk factor for frontotemporal lobar degeneration (FTLD), a group of clinically, pathologically and genetically heterogeneous disorders associated with atrophy in the frontal lobe and temporal lobe of the brain. The actual role of TMEM106B, and that of the closely related protein TMEM106A are still undetermined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM106B
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene TMEM106B
Gene Accession No. Q9NUM4
Gene Alias 2310036D22Rik; 5830455K21Rik; 6430519M21Rik; AI428776; AI661344; hypothetical protein LOC508903; LRRGT00101; tmem106b; tmem106bb; Transmembrane protein 106B; transmembrane protein 106Bb; zgc:114147
Gene Symbols TMEM106B
Host Species Rabbit
Immunogen A 15 amino acid synthetic peptide near the amino terminus of human TMEM106B.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54664
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.