Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM106B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563558
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563558 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563558 Supplier Invitrogen™ Supplier No. PA563558
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GKSLSHLPLH SSKEDAYDGV TSENMRNGLV NSEVHNEDGR NGDVSQFPYV EF Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 87%.

Transmembrane protein 106B (TMEM106B) is a single-pass transmembrane protein that is thought to be a novel risk factor for frontotemporal lobar degeneration (FTLD), a group of clinically, pathologically and genetically heterogeneous disorders associated with atrophy in the frontal lobe and temporal lobe of the brain. The actual role of TMEM106B, and that of the closely related protein TMEM106A are still undetermined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM106B
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TMEM106B
Gene Accession No. Q9NUM4
Gene Alias 2310036D22Rik; 5830455K21Rik; 6430519M21Rik; AI428776; AI661344; hypothetical protein LOC508903; LRRGT00101; tmem106b; tmem106bb; Transmembrane protein 106B; transmembrane protein 106Bb; zgc:114147
Gene Symbols TMEM106B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TMEM106B. Recombinant protein control fragment (Product #RP-100479).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54664
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.