Learn More
TMEM106B Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303439AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
Transmembrane protein 106B (TMEM106B) is a single-pass transmembrane protein that is thought to be a novel risk factor for frontotemporal lobar degeneration (FTLD), a group of clinically, pathologically and genetically heterogeneous disorders associated with atrophy in the frontal lobe and temporal lobe of the brain. The actual role of TMEM106B, and that of the closely related protein TMEM106A are still undetermined.Specifications
TMEM106B | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q80X71, Q9NUM4 | |
TMEM106B | |
Between 1 and 50 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
TMEM106B | |
TMEM106B, Transmembrane protein 106B | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54664, 71900 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.