Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMEM106B Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981589
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1589 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1589 Supplier Bioss Supplier No. BS11694R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Transmembrane protein 106B (TMEM106B) is a single-pass transmembrane protein that is thought to be a novel risk factor for frontotemporal lobar degeneration (FTLD), a group of clinically, pathologically and genetically heterogeneous disorders associated with atrophy in the frontal lobe and temporal lobe of the brain. The actual role of TMEM106B, and that of the closely related protein TMEM106A are still undetermined.

Specifications

Antigen TMEM106B
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene TMEM106B
Gene Accession No. Q9NUM4
Gene Alias 2310036D22Rik; 5830455K21Rik; 6430519M21Rik; AI428776; AI661344; BOS_4052; LOC100356191; LRRGT00101; Tmem106b; tmem106bb; Transmembrane protein 106B; transmembrane protein 106Bb; transmembrane protein 106C-like; zgc:114147
Gene Symbols TMEM106B
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human TMEM106B .
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54664
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.