Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM106C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104354
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104354 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104354 Supplier Invitrogen™ Supplier No. PA5104354
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total T106C.

TMEM106C is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM106C
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TMEM106C
Gene Accession No. Q5RJK0, Q80VP8, Q9BVX2
Gene Alias AI046681; BC046621; D15Ertd405e; EMOC; Endoplasmic reticulum membrane protein overexpressed in cancer; hypothetical protein LOC514271; RGD1311532; TMEM106C; Transmembrane protein 106C; transmembrane protein 106C-like protein
Gene Symbols TMEM106C
Host Species Rabbit
Immunogen A synthesized peptide derived from human TMEM106C(Accession Q9BVX2), corresponding to amino acid residues G56-L106.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 315286, 380967, 79022
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.