Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TMEM108 Antibody, Biotin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP321447B
Description
TMEM108 Polyclonal antibody specifically detects TMEM108 in Human samples. It is validated for ImmunofluorescenceSpecifications
TMEM108 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence | |
cancer/testis antigen 124, CT124, KIAA1690, MGC3040, transmembrane protein 108 | |
Produced in rabbits immunized with E. coli-derived Human TMEM108 fragment | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunofluorescence | |
Biotin | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
66000 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction