Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM109 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112198
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112198 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112198 Supplier Invitrogen™ Supplier No. PA5112198
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TMEM109 Polyclonal Antibody for Western Blot, IP

May mediate cellular response to DNA damage by protecting against ultraviolet C-induced cell death (PubMed:23542032). Can form voltage-gated calcium and potassium channels in vitro. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM109
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.03% ProClin 300; pH 7
Gene TMEM109
Gene Accession No. Q9BVC6
Gene Alias 1110006I15Rik; AA958958; C77652; hypothetical LOC540138; mg23; MGC5508; mitsugumin 23; mitsugumin23; mitsugumin-23; TMEM109; Transmembrane protein 109
Gene Symbols TMEM109
Host Species Rabbit
Immunogen E. coli-derived Human TMEM109 fragment.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79073
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.