Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TMEM109 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5116051
Description
Antibody detects endogenous levels of total TMEM109.
May mediate cellular response to DNA damage by protecting against ultraviolet C-induced cell death (PubMed:23542032). Can form voltage-gated calcium and potassium channels in vitro. [UniProt].
Specifications
| TMEM109 | |
| Polyclonal | |
| Unconjugated | |
| TMEM109 | |
| 1110006I15Rik; AA958958; C77652; hypothetical LOC540138; mg23; MGC5508; mitsugumin 23; mitsugumin23; mitsugumin-23; TMEM109; Transmembrane protein 109 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 361732, 79073 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| Q6AYQ4, Q9BVC6 | |
| TMEM109 | |
| A synthesized peptide derived from human TMEM109(Accession Q9BVC6), corresponding to amino acid residues K16-D66. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction