Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100354
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100354 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100354 Supplier Invitrogen™ Supplier No. PA5100354
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total TMEM11.

Plays a role in mitochondrial morphogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM11
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TMEM11
Gene Accession No. B0BN86, P17152, Q8BK08
Gene Alias 5730466P16Rik; AA409091; C17orf35; PM1; PMI; Protein PM1; Protein PMI; putative receptor protein; RGD1309817; TMEM11; transmembrane protein 11; transmembrane protein 11, mitochondrial
Gene Symbols TMEM11
Host Species Rabbit
Immunogen A synthesized peptide derived from human TMEM11(Accession P17152), corresponding to amino acid residues C119-R169.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 216821, 303196, 8834
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.