Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TMEM114 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570847
Description
This target displays homology in the following species: Cow: 92%; Human: 100%; Mouse: 92%; Pig: 92%; Rat: 85%.
TMEM114 is a protein coding gene.
Specifications
TMEM114 | |
Polyclonal | |
Unconjugated | |
TMEM114 | |
4930511J11Rik; claudin 26; claudin-26; Cldn26; RGD1560593; TMEM114; Transmembrane protein 114 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
283953 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
B3SHH9 | |
TMEM114 | |
synthetic peptide directed towards the N-terminal region of human TMEM114. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction