Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TMEM115 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP169572
Description
TMEM115 Polyclonal specifically detects TMEM115 in Human samples. It is validated for Western Blot.Specifications
TMEM115 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
PL6Placental protein 6, PP6, Protein PL6, transmembrane protein 115 | |
Rabbit | |
38 kDa | |
100 μL | |
Primary | |
Zebrafish: 85%. | |
Human, Mouse, Rat, Bovine, Canine, Guinea Pig, Rabbit, Zebrafish | |
IgG |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
Q12893 | |
TMEM115 | |
Synthetic peptides corresponding to TMEM115(transmembrane protein 115) The peptide sequence was selected from the N terminal of TMEM115. Peptide sequence LLSFAVDTGCLAVTPGYLFPPNFWIWTLATHGLMEQHVWDVAISLTTVVV. | |
Affinity purified | |
RUO | |
11070 | |
Centrifuge prior to opening. Reconstitute with sterilized PBS to a final concentration of 1mg/ml. Vortex followed by Centrifuge again to pellet the solution. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction