Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM115 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542929
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542929 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542929 Supplier Invitrogen™ Supplier No. PA542929
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: LLSFAVDTGC LAVTPGYLFP PNFWIWTLAT HGLMEQHVWD VAISLTTVVV Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 85%.

The specific function of TMEM115 is not yet known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM115
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TMEM115
Gene Accession No. Q12893
Gene Alias C78915; hypothetical protein LOC619264; LUCA11.2; PL6; PL6 protein; placental protein 6; Pp6; Protein PL6; Protein PL6 homolog; RGD1310540; testicular tissue protein Li 201; TMEM115; Transmembrane protein 115; wu:fc43c03; wu:fd18h12; zgc:114074; zgc:114074 protein
Gene Symbols TMEM115
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human TMEM115 (aa 35-84).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11070
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.