Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM118 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564157
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564157 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564157 Supplier Invitrogen™ Supplier No. PA564157
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QPHHHFHHGG HRGGSLLQHV GGDHRGHSEE GGDEQPGTPA PALSELKAVI CWLQKGLP Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 95%.

TMEM118 is a multi-pass membrane protein, and contains 1 RING-type zinc finger. The function of TMEM118 remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM118
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RNFT2
Gene Accession No. Q96EX2
Gene Alias AW049082; B830028P19Rik; hypothetical protein LOC553554; RGD1560195; RING finger and transmembrane domain-containing protein 2; RING finger and transmembrane domain-containing protein 2; ring finger protein, transmembrane 2; ring finger protein, transmembrane 2; RNFT2; si:dkey-175n1.1; Tmem118; Transmembrane protein 118; zgc:109947
Gene Symbols RNFT2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TMEM118. Recombinant protein control fragment (Product #RP-102010).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84900
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.