Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM118 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543500
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543500 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543500 Supplier Invitrogen™ Supplier No. PA543500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: HGGHRGGSLL QHVGGDHRGH SEEGGDEQPG TPAPALSELK AVICWLQKGL Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 79%.

TMEM118 is a multi-pass membrane protein, and contains 1 RING-type zinc finger. The function of TMEM118 remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM118
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene RNFT2
Gene Accession No. Q96EX2
Gene Alias AW049082; B830028P19Rik; hypothetical protein LOC553554; RGD1560195; RING finger and transmembrane domain-containing protein 2; RING finger and transmembrane domain-containing protein 2; ring finger protein, transmembrane 2; ring finger protein, transmembrane 2; RNFT2; si:dkey-175n1.1; Tmem118; Transmembrane protein 118; zgc:109947
Gene Symbols RNFT2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human TMEM118 (aa 118-167).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84900
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.