Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TMEM119 Antibody (2699A), PE/Cy7, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB103131PECY7
Description
TMEM119 Monoclonal antibody specifically detects TMEM119 in Human samples. It is validated for Flow CytometrySpecifications
TMEM119 | |
Monoclonal | |
PE-Cyanine7 | |
PBS | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
RUO | |
338773 | |
Store at 4C in the dark. Do not freeze. | |
IgG |
Flow Cytometry | |
2699A | |
Flow Cytometry | |
OBIF, Osteoblast Induction Factor, TMEM119 | |
Chinese Hamster Ovary cell line CHO derived human TREM119, Arg26-Met96, Accession # Q4V9L6.1 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction