Learn More
Invitrogen™ TMEM119 Monoclonal Antibody (CL8714)

Description
Immunogen sequence: GDGARMVEGR GAEEEEKGSQ EGDQEVQGHG VPVETPEAQE EPCSGVLEGA VVAGEGQGEL EGSLLLAQEA QGPVGPPESP CACSSVHPS Highest antigen sequence indentity to the following orthologs: Mouse - 52%, Rat - 46%.
Specifications
Specifications
| Antigen | TMEM119 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | CL8714 |
| Concentration | 1.00 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | TMEM119 |
| Gene Accession No. | Q4V9L6 |
| Gene Alias | AW208946; BC025600; OBIF; Osteoblast induction factor; PSEC0199; Tmem119; Transmembrane protein 119; UNQ731/PRO1415 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.