Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM123 Monoclonal Antibody (297617)
GREENER_CHOICE

Catalog No. PIMA524007
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524007 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524007 Supplier Invitrogen™ Supplier No. MA524007
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute at 0.5 mg/mL in sterile PBS.

Transmembrane protein 123, or Tmem123, is a single-pass type I membrane protein and belongs to the CD164 family. Tmem123 acts as a novel marker for DC maturation. DCs are also known as professional antigen-presenting cells. It is reported that the Tmem123 molecule may be closely associated with the cell surface expression of CD40. It was also reported previously that Tmem123 protein may also be responsible for oncotic cell death, which is characterized by cell swelling, organelle swelling, vacuolization, increased membrane permeability and cell adhesion.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM123
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone 297617
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene TMEM123
Gene Accession No. Q8N131
Gene Alias 2310075C12Rik; KCT3; KCT-3; keratinocytes associated transmembrane protein 3; keratinocytes-associated transmembrane protein 3; Porimin; PORMIN; pro oncosis receptor inducing membrane injury; pro-oncosis receptor inducing membrane injury; PSEC0111; RGD1305625; serine/threonine-rich receptor; TMEM123; Transmembrane protein 123; UNQ641/PRO1271
Gene Symbols TMEM123
Host Species Mouse
Immunogen CHO-derived recombinant human Porimin His27-Ser149.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114908
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.