Learn More
Invitrogen™ TMEM123 Polyclonal Antibody

Description
The amino acid sequence used as immunogen is 94% in White-cheeked Gibbon, and 88% homologous in Equine, and Chicken. Immunogen sequence: SRRGIRYRSI DEHDAII.
Specifications
Specifications
| Antigen | TMEM123 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA and 0.05% sodium azide |
| Gene | TMEM123 |
| Gene Accession No. | Q5HZB0 |
| Gene Alias | 2310075C12Rik; KCT3; KCT-3; keratinocytes associated transmembrane protein 3; keratinocytes-associated transmembrane protein 3; Porimin; PORMIN; pro oncosis receptor inducing membrane injury; pro-oncosis receptor inducing membrane injury; PSEC0111; RGD1305625; serine/threonine-rich receptor; TMEM123; Transmembrane protein 123; UNQ641/PRO1271 |
| Gene Symbols | TMEM123 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.