Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM123 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA129811
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA129811 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA129811 Supplier Invitrogen™ Supplier No. PA129811
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA1-29811 detects Porimin from bovine, mouse, human, rat samples.

Transmembrane protein 123, or Tmem123, is a single-pass type I membrane protein and belongs to the CD164 family. Tmem123 acts as a novel marker for DC maturation. DCs are also known as professional antigen-presenting cells. It is reported that the Tmem123 molecule may be closely associated with the cell surface expression of CD40. It was also reported previously that Tmem123 protein may also be responsible for oncotic cell death, which is characterized by cell swelling, organelle swelling, vacuolization, increased membrane permeability and cell adhesion.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM123
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA and 0.05% sodium azide
Gene TMEM123
Gene Accession No. Q8N131, Q91Z22
Gene Alias 2310075C12Rik; KCT3; KCT-3; keratinocytes associated transmembrane protein 3; keratinocytes-associated transmembrane protein 3; Porimin; PORMIN; pro oncosis receptor inducing membrane injury; pro-oncosis receptor inducing membrane injury; PSEC0111; RGD1305625; serine/threonine-rich receptor; TMEM123; Transmembrane protein 123; UNQ641/PRO1271
Gene Symbols TMEM123
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 173-188 SRRGIRYRTIDEHDAI of human Porimin.
Purification Method Protein G
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114908, 71929
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.