Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMEM126B Polyclonal Antibody, Invitrogen™

Catalog No. PIPA553178
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553178 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553178 Supplier Thermo Scientific Supplier No. PA553178

Rabbit Polyclonal Antibody

Immunogen sequence: IQVWGVGTEE APKVFKMAAS MHGQPSPSLE DAKLRRPMVI EIIE Highest antigen sequence identity to the following orthologs: Mouse - 50%, Rat - 45%.

The function remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM126B
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TMEM126B
Gene Accession No. Q8IUX1
Gene Alias 1110001A23Rik; complex I assembly factor TMEM126B, mitochondrial; HT007; RGD1308371; TMEM126B; Transmembrane protein 126B
Gene Symbols TMEM126B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TMEM126B
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55863
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.