Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM127 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144339
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144339 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144339 Supplier Invitrogen™ Supplier No. PA5144339
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TMEM127 Polyclonal Antibody for ELISA, ICC/IF, IHC (P)

TMEM127 controls cell proliferation acting as a negative regulator of TOR signaling pathway mediated by mTORC1. May act as a tumor suppressor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM127
Applications ELISA, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 3.35 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene TMEM127
Gene Accession No. O75204
Gene Alias 2310003P10Rik; AI314202; AI317350; hypothetical protein LOC553545; RGD1309744; si:dkey-49h9.5; tmem127; Transmembrane protein 127; zgc:109899
Gene Symbols TMEM127
Host Species Rabbit
Immunogen Recombinant Human Transmembrane protein 127 protein (1-95AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55654
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.