Learn More
TMEM127 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303450AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
TMEM127 controls cell proliferation acting as a negative regulator of TOR signaling pathway mediated by mTORC1. May act as a tumor suppressor.Specifications
TMEM127 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
O75204, Q8BGP5 | |
TMEM127 | |
Between 1 and 50 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
TMEM127 | |
TMEM127 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55654, 69470 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.