Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TMEM131 Antibody, DyLight 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP121369G
Description
TMEM131 Polyclonal antibody specifically detects TMEM131 in Human samples. It is validated for Western BlotSpecifications
TMEM131 | |
Polyclonal | |
Western Blot | |
KIAA02572610524E03Rik, PRO1048, Protein RW1, RW1CC28, transmembrane protein 131, YR-23 | |
The immunogen recognized by this antibody maps to a region between residue 1779 and 1829 of human transmembrane protein 131 using the numbering given in entry BAA13387.1 (GeneID 23505). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
DyLight 488 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
23505 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction