Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMEM131 Polyclonal Antibody, Bethyl Laboratories

Rabbit Polyclonal Antibody

$151.31

Specifications

Antigen TMEM131
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Form Liquid
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-156-1847
Encompass_Preferred
View Documents
Bethyl Laboratories, Inc
A302129AT
20 μL
Each of 1 for $151.31
Only null left
Add to Cart
 
Description

Description

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse

Inhibitor of Nuclear factor Kappa-B kinase subunit alpha (IKK alpha) is encoded by the CHUK gene and located on chromosome 10 in humans. It is also known as IKK1 or CHUK (Conserved helix-loop-helix ubiquitous kinase). IKK alpha belongs to the family of serine threonine protein kinase and forms a part of IKK complex. The IKK complex is required for the activation of NF-kB with IKK alpha specifically known to be involved in the non-canocial pathway for NF-kB activation. IKK alpha along with IKK beta and a regulatory protein NEMO (IKK gamma) phosphorylates IkB proteins which targets the later for ubiquitin mediated degradation. It is involved with several cellular processes with extensive studies on its role in inflammation and cell survival.
Specifications

Specifications

TMEM131
Polyclonal
Liquid
RUO
TBS with 0.1% BSA and 0.09% sodium azide
2610524E03Rik, CC28, KIAA0257, PRO1048, Protein RW1, RW1, YR-23
TMEM131
IgG
Antigen affinity chromatography
Antibody
Western Blot
Unconjugated
Rabbit
Human
Q92545
23505
Between 1779 and 1829
Primary
4° C
TMEM131
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.