Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM132A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569464
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569464 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569464 Supplier Invitrogen™ Supplier No. PA569464
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Pig: 93%; Rabbit: 92%; Rat: 100%.

This gene encodes a protein that is highly similar to the rat Grp78-binding protein (GBP). Alternatively spliced transcript variants encoding different isoforms have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM132A
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TMEM132A
Gene Accession No. Q24JP5
Gene Alias 6720481D13Rik; GBP; glucose-regulated protein, 78kDa; GRP78 binding protein; GRP78-binding protein; heat shock 70kDa protein 5 (glucose-regulated protein, 78kDa) binding protein 1; heat shock 70kDa protein 5 binding protein 1; heat shock protein 5 binding protein 1; HSPA5-binding protein 1; Hspa5bp1; KIAA1583; LOW QUALITY PROTEIN: transmembrane protein 132A; Orai1; R74613; TMEM132A; Transmembrane protein 132A
Gene Symbols TMEM132A
Host Species Rabbit
Immunogen synthetic peptide directed towards the N-terminal region of human TMEM132A.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54972
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.