Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TMEM132A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA585903
Description
TMEM132A Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
This gene encodes a protein that is highly similar to the rat Grp78-binding protein (GBP). Alternatively spliced transcript variants encoding different isoforms have been described.
Specifications
TMEM132A | |
Polyclonal | |
Unconjugated | |
TMEM132A | |
6720481D13Rik; GBP; glucose-regulated protein, 78kDa; GRP78 binding protein; GRP78-binding protein; heat shock 70kDa protein 5 (glucose-regulated protein, 78kDa) binding protein 1; heat shock 70kDa protein 5 binding protein 1; heat shock protein 5 binding protein 1; HSPA5-binding protein 1; Hspa5bp1; KIAA1583; LOW QUALITY PROTEIN: transmembrane protein 132A; Orai1; R74613; TMEM132A; Transmembrane protein 132A | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54972, 98170 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 0.2% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4 | |
Q24JP5, Q922P8 | |
TMEM132A | |
Synthetic peptide within Human Transmembrane protein 132A aa 204-253. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction