Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMEM132B Polyclonal Antibody, Invitrogen™

Catalog No. PIPA569678
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569678 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569678 Supplier Thermo Scientific Supplier No. PA569678

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 86%; Dog: 86%; Guinea Pig: 77%; Horse: 93%; Human: 100%; Mouse: 79%

TMEM132B is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM132B
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TMEM132B
Gene Accession No. Q14DG7
Gene Alias AK220418; KIAA1786; KIAA1906; mKIAA1786; TMEM132B; Transmembrane protein 132B
Gene Symbols TMEM132B
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human TMEM132B
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114795
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.