Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM132D Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595702
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595702 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595702 Supplier Invitrogen™ Supplier No. PA595702
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DFSLMATSLD EKVVSIHQDP KFKWPIIAAE TEGQGTLVKV EMVISESCQK SKRKSVLAVG TANIKVKFGQ NDANPNTSDS RHTGAGVHME NNVSDRRPKK PSQEWGSQEG QYYGSSSMGL MEGRGTTTDR SILQKKKGQE SLLDDNSHLQ TIPSDLTSFP AQVDLPRSNG EMDGNDLMQA SKGLSD; Positive Samples: U-251MG, Mouse brain, Mouse liver, Rat brain; Cellular Location: Membrane, Single-pass type I membrane protein.

TMEM132D may serve as a cell-surface marker for oligodendrocyte differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM132D
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.02 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene TMEM132D
Gene Accession No. Q14C87, Q76HP2, Q76HP3
Gene Alias C630028F04Rik; HBE120; hypothetical protein; KIAA1944; Mature OL transmembrane protein; mature oligodendrocyte transmembrane protein; mature oligodendrocytes transmembrane protein; MOLT; PPP1R153; protein phosphatase 1, regulatory subunit 153; TMEM132D; Transmembrane protein 132D
Gene Symbols TMEM132D
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 730-915 of human TMEM132D (NP_5977052).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 121256, 243274, 288750
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.