Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM132D Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA523522
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA523522 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA523522 Supplier Invitrogen™ Supplier No. PA523522
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TMEM132D Polyclonal Antibody for Western Blot, Flow

TMEM132D may serve as a cell-surface marker for oligodendrocyte differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM132D
Applications Flow Cytometry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene TMEM132D
Gene Accession No. Q14C87
Gene Alias C630028F04Rik; HBE120; hypothetical protein; KIAA1944; Mature OL transmembrane protein; mature oligodendrocyte transmembrane protein; mature oligodendrocytes transmembrane protein; MOLT; PPP1R153; protein phosphatase 1, regulatory subunit 153; TMEM132D; Transmembrane protein 132D
Gene Symbols TMEM132D
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 805-833 amino acids from the C-terminal region of human TMEM132D.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 121256
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.