Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TMEM132E Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5141158
This item is not returnable.
View return policy
Description
TMEM132E Polyclonal Antibody for Western Blot
Required for normal inner ear hair cell function and hearing.
Specifications
| TMEM132E | |
| Polyclonal | |
| Unconjugated | |
| TMEM132E | |
| DFNB99; Gm644; TMEM132E; Transmembrane protein 132E | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 124842, 270893 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 0.2% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q6IEE6, Q6IEE7 | |
| TMEM132E | |
| Synthetic peptide within Human TMEM132E aa 10-59/1,074. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction