Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TMEM139 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | TMEM139 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
TMEM139 Polyclonal specifically detects TMEM139 in Human samples. It is validated for Western Blot.Specifications
| TMEM139 | |
| Polyclonal | |
| Rabbit | |
| Q8IV31 | |
| 135932 | |
| Synthetic peptides corresponding to TMEM139 (transmembrane protein 139) The peptide sequence was selected from the N terminal of TMEM139. Peptide sequence ITPVAYFFLTLGGFFLFAYLLVRFLEWGLRSQLQSMQTESPGPSGNARDN. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| FLJ90586, transmembrane protein 139 | |
| TMEM139 | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title