Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TMEM140 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144108
Description
TMEM140 Polyclonal Antibody for ICC/IF, IHC (P), ELISA
TMEM140 is a gene that encodes a protein with unknown function. It is located on the long arm of chromosome 17 in humans and is conserved across different species. While the specific function of TMEM140 is not well understood, studies have suggested that it may be involved in the regulation of cell proliferation and differentiation.
Specifications
TMEM140 | |
Polyclonal | |
Unconjugated | |
TMEM140 | |
TMEM140; Transmembrane protein 140 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55281 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
Q9NV12 | |
TMEM140 | |
Recombinant Human Transmembrane protein 140 protein (33-77AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction