Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TMEM145 Polyclonal Antibody
TMEM145 Polyclonal Antibody
Supplier: Thermo Scientific PA566327
Description
TMEM145 Polyclonal Antibody for ICC/IF, IHC

Specifications
TMEM145 | |
Polyclonal | |
Unconjugated | |
TMEM145 | |
B930076A02Rik; Tmem145; Transmembrane protein 145 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
284339 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NBT3 | |
TMEM145 | |
Recombinant Human TMEM145. Recombinant protein control fragment (Product #RP-105152). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction