Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    TMEM147 Antibody, mFluor Violet 500 SE, Novus Biologicals™
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP282028MFV500
Description
TMEM147 Polyclonal antibody specifically detects TMEM147 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISASpecifications
| TMEM147 | |
| Polyclonal | |
| Western Blot, ELISA | |
| MGC1936, NIFIE14, Protein NIFIE 14, seven transmembrane domain protein, transmembrane protein 147 | |
| TMEM147 antibody was raised against a 13 amino acid synthetic peptide near the center of human TMEM147. The immunogen is located within amino acids 70 - 120 of TMEM147. Amino Acid Squence: AAMSRNAGKGEYKIM | |
| 0.1 mL | |
| Cell Biology, Neuroscience | |
| 10430 | |
| Store at 4C in the dark. | |
| IgG | 
| Western Blot, ELISA | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction