Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TMEM147 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA572696
Description
TMEM147 Polyclonal Antibody for Western Blot, ELISA
TMEM147 is a protein coding gene.
Specifications
TMEM147 | |
Polyclonal | |
Unconjugated | |
TMEM147 | |
2010004E11Rik; 5033425B17Rik; hypothetical protein LOC535038; Nifie14; Protein NIFIE 14; RGD1304706; seven transmembrane domain protein; TMEM147; Transmembrane protein 147 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10430, 292792, 69804 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q2TA63, Q9BVK8, Q9CQG6 | |
TMEM147 | |
TMEM147 antibody for a 13 amino acid synthetic peptide near the center of human TMEM147. The immunogen is within amino acids 70 - 120 of TMEM147. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction