Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM147 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595876
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595876 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595876 Supplier Invitrogen™ Supplier No. PA595876
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GLNLVMSRNA GKGEYK.

TMEM147 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM147
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.35 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene TMEM147
Gene Accession No. Q9BVK8, Q9CQG6
Gene Alias 2010004E11Rik; 5033425B17Rik; hypothetical protein LOC535038; Nifie14; Protein NIFIE 14; RGD1304706; seven transmembrane domain protein; TMEM147; Transmembrane protein 147
Gene Symbols TMEM147
Host Species Rabbit
Immunogen Synthetic peptide
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10430, 69804
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.