Learn More
Invitrogen™ TMEM147 Polyclonal Antibody

Description
Recommended positive controls: Raji. Predicted reactivity: Mouse (100%), Rat (100%), Zebrafish (100%), Xenopus laevis (100%), Rhesus Monkey (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | TMEM147 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 |
| Gene | TMEM147 |
| Gene Accession No. | Q9BVK8 |
| Gene Alias | 2010004E11Rik; 5033425B17Rik; hypothetical protein LOC535038; Nifie14; Protein NIFIE 14; RGD1304706; seven transmembrane domain protein; TMEM147; Transmembrane protein 147 |
| Gene Symbols | TMEM147 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.