Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM14A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567246
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567246 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567246 Supplier Invitrogen™ Supplier No. PA567246
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AGYGAYRVSND KRDVKVSLF Highest antigen sequence identity to the following orthologs - mouse 95%, rat 100%.

Inhibits apoptosis via negative regulation of the mitochondrial outer membrane permeabilization involved in apoptotic signaling pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM14A
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TMEM14A
Gene Accession No. Q9Y6G1
Gene Alias C6orf73; PTD011; TMEM14A; Transmembrane protein 14A
Gene Symbols TMEM14A
Host Species Rabbit
Immunogen Recombinant Human TMEM14A. Recombinant protein control fragment (Product #RP-107885).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28978
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.