Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TMEM14B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5112746
Description
TMEM14B Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

Specifications
TMEM14B | |
Polyclonal | |
Unconjugated | |
TMEM14B | |
TMEM14B; Transmembrane protein 14B | |
Rabbit | |
Protein G | |
RUO | |
81853 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
4.5 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q9NUH8 | |
TMEM14B | |
Recombinant Human Transmembrane protein 14B protein (1-114AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction