Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM150C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562704
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562704 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562704 Supplier Invitrogen™ Supplier No. PA562704
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LPLNSAERKP GVKHAPYISI AGDDPPAS Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 89%.

This gene encodes a transmembrane protein component of a mechanosensitve ion channel that is activated by mechanical stimuli in various cell types and confers slowly adapting, mechanically activated currents in dorsal root ganglion neurons. Mechanically activated ion channels are sensors that are critical for hearing, touch, pain, and blood pressure regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM150C
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TMEM150C
Gene Accession No. B9EJG8
Gene Alias gb:dn898596; RGD1306105; si:ch211-249h16.6; Tentonin 3; TMEM150C; Transmembrane protein 150C; Ttn3
Gene Symbols TMEM150C
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TMEM150C. Recombinant protein control fragment (Product #RP-100677).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 441027
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.