Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TMEM156 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306638JF646
Description
TMEM156 Polyclonal antibody specifically detects TMEM156 in Human samples. It is validated for ELISASpecifications
| TMEM156 | |
| Polyclonal | |
| ELISA | |
| FLJ23235, transmembrane protein 156 | |
| Produced in rabbits immunized with purified, recombinant Human TMEM156 (Accession#: AAH30803.1; Lys26-Ser211) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 80008 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction