Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    TMEM156 Antibody, mFluor Violet 500 SE, Novus Biologicals™
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306638MFV500
Description
TMEM156 Polyclonal antibody specifically detects TMEM156 in Human samples. It is validated for ELISASpecifications
| TMEM156 | |
| Polyclonal | |
| ELISA | |
| FLJ23235, transmembrane protein 156 | |
| Produced in rabbits immunized with purified, recombinant Human TMEM156 (Accession#: AAH30803.1; Lys26-Ser211) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified | 
| ELISA | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 80008 | |
| Store at 4C in the dark. | |
| IgG | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction