Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM158 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100204
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100204 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100204 Supplier Invitrogen™ Supplier No. PA5100204
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total TM158.

Receptor for brain injury-derived neurotrophic peptide (BINP), a synthetic 13-mer peptide.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM158
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TMEM158
Gene Accession No. Q6F5E0, Q8WZ71
Gene Alias 2310037P21Rik; 40 kDa BINP-binding protein; BBP; BINP binding protein p40BBP; BINP receptor; brain injury-derived neurotrophic peptide (BINP) binding protein; brain specific binding protein; brain-specific binding protein; HBBP; Mbbp; p40BBP; Ras induced senescence 1; Ras-induced senescence 1; ras-induced senescence protein 1; Ris1; Tmem158; Transmembrane protein 158; transmembrane protein 158 (gene/pseudogene)
Gene Symbols TMEM158
Host Species Rabbit
Immunogen A synthesized peptide derived from human TMEM158(Accession Q8WZ71), corresponding to amino acid residues P184-V234.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25907, 72309
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.