Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMEM158 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981654
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1654 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1654 Supplier Bioss Supplier No. BS11770R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Receptor for brain injury-derived neurotrophic peptide (BINP), a synthetic 13-mer peptide.

Specifications

Antigen TMEM158
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene TMEM158
Gene Accession No. Q8WZ71
Gene Alias 2310037P21Rik; 40 kDa BINP-binding protein; BBP; BINP binding protein p40BBP; BINP receptor; brain injury-derived neurotrophic peptide (BINP) binding protein; brain specific binding protein; HBBP; Mbbp; p40BBP; Ras induced senescence 1; Ras-induced senescence 1; Ras-induced senescence protein 1; RIS1; TMEM158; Transmembrane protein 158; transmembrane protein 158 (gene/pseudogene)
Gene Symbols TMEM158
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human TMEM158.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25907
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.