Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM161A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543020
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543020 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543020 Supplier Invitrogen™ Supplier No. PA543020
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: LLAMLVQVVR EETLELGLEP GLASMTQNLE PLLKKQGWDW ALPVAKLAIR Sequence homology: Cow: 100%; Dog: 93%; Guinea Pig: 86%; Horse: 100%; Human: 100%; Mouse: 100%; Rat: 100%.

May play a role in protection against oxidative stress. Overexpression leads to reduced levels of oxidant-induced DNA damage and apoptosis. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM161A
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TMEM161A
Gene Accession No. Q9NX61
Gene Alias adaptive response to oxidative stress protein 29; AI428876; AROS-29; BB161850; BC021367; RGD1307703; TMEM161A; Transmembrane protein 161A; UNQ582/PRO1152
Gene Symbols TMEM161A
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human TMEM161A.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54929
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.