Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM161A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598196
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598196 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598196 Supplier Invitrogen™ Supplier No. PA598196
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TMEM161A Polyclonal Antibody for ELISA, ICC/IF, IHC (P), Western Blot

May play a role in protection against oxidative stress. Overexpression leads to reduced levels of oxidant-induced DNA damage and apoptosis. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM161A
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene TMEM161A
Gene Accession No. Q9NX61
Gene Alias adaptive response to oxidative stress protein 29; AI428876; AROS-29; BB161850; BC021367; RGD1307703; TMEM161A; Transmembrane protein 161A; UNQ582/PRO1152
Gene Symbols TMEM161A
Host Species Rabbit
Immunogen Recombinant Human Transmembrane protein 161A protein (29-98AA).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54929
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.