Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM165 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111548
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111548 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111548 Supplier Invitrogen™ Supplier No. PA5111548
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DKTFFIAAIM AMRYNRLTVL Highest antigen sequence indentity to the following orthologs: Rat - 100%, Mouse - 100%.

The function of TM165 remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM165
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TMEM165
Gene Accession No. Q9HC07
Gene Alias AV026557; CDG2K; FT27; GDT1; pFT27; pt27; tm165; Tmem165; TMPT27; TPA regulated locus; Tpardl; TPA-regulated locus protein; Tparl; transmembrane protein 164; transmembrane protein 165; Transmembrane protein PFT27; transmembrane protein PT27; Transmembrane protein TPARL
Gene Symbols TMEM165
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human TMEM165. Recombinant protein control fragment (Product #RP-108470).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55858
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.